A major plus for routine analysis
New Spectroquant® Prove plus spectrophotometers with added speed, storage, and simplicity
As your trusted partner in analytical chemistry, we’re always working on the next innovation to enhance your analytical work. Our latest generation of spectrophotometers for your analysis does just that. The Spectroquant® Prove plus range is packed with features that make accuracy and precision simpler than ever.
Positives of Prove plus for water, environmental and food & beverage analysis
- Smoother functionality with an instantly responsive display and faster processing
- Greater capacity to store more measurement results (up to 7000)
- Extended warranty period of two years
Demo/Angebot anfordern
Features and Benefits
Pre-programmed to work seamlessly with more than 150 Spectroquant® test kits, Spectroquant® Prove plus photometers are among the most trusted for your analysis.
- Easy to use and pre-programmed with pre-defined reagents
- Reduced use of chemicals compared to traditional methods
- Suitable for a broad measuring range per parameter (minimizes dilution work)
- Pre-programmed for over 300 analysis methods
- Cell size up to 10 cm with Prove 600 plus for high-sensitivity measurements
Spectroquant® PROVE 100 | Spectroquant® PROVE 300 | Spectroquant® PROVE 600 |
Prove 100 plus | Prove 300 plus | Prove 600 plus |
Routine VIS measurements in wastewater analysis and environmental testing Examples: Groundwater, effluents, soil |
Sensitive UV/VIS analysis of drinking water, food and beverages Examples: Drinking water, beverages, special beer applications, sugar, olive oil |
Complex UV/Vis measurements in process water testing Examples: Power plants, cooler & boiler water |
1.73026.0001 | 1.73027.0001 | 1.73028.0001 |
Einleitung
So starten Sie das Prove
So navigieren Sie durch die Funktionen
So verwenden Sie das Prove mit den Spectroquant® Zelltestkits und Reagenzien-Sets
So reinigen Sie das Prove auf einfache Weise
So wählen Sie Sprache, Datum und Uhrzeit aus
Datentransfer – Daten auf Anfrage – einfache Filterung – einaches Teilen
So prüfen Sie auf eine störende Matrix
Gewährleisten Sie die Genauigkeit Ihres Systems: Verwenden Sie AQA Analytical Quality Assurance