Try Polyplus-transfection® reagents in your CRISPR experiments
Polyplus-transfection® SA is a biotechnology company that develops and sells innovative solutions for the delivery of nucleic acids in research, bioproduction and therapeutics. Polyplus-transfection® is ISO 9001-certified since 2002 and supplies, through VWR International, its wide range of reagents for the transfection of nucleic acids, such as DNA, mRNA, oligonucleotides and siRNA.
Its complete range of transfection reagents for CRISPR application enables researchers to find adapted product(s) for their experimental approach(es). There are three main systems to transfect guide RNA and express Cas9 protein which are DNA, RNA or RNP (gRNA + Cas9 protein) approaches. Each method will be performed with a specific reagent:
- DNA approach: Use jetPRIME®
- RNA approach: Use jetMESSENGER®
- RNP approach: Use jetCRISPR™ + SpCas9 Nuclease
Besides, Polyplus-transfection® also provides technical support services to help customers to optimize their transfection (94% of satisfied customers).
We now provide a full range of reagents for all your CRISPR experiments:
Delivered Molecule | Cas9 | Guide RNA | Polyplus Solution |
DNA | Plasmid | Plasmid | jetPRIME® |
DNA/RNA | Plasmid | RNA | jetPRIME® |
RNA/RNA | mRNA | RNA | jetMESSENGER™ |
RNA/protein (RNP) | Protein | RNA | jetCRISPR™ |
jetCRISPR™ - RNP transfection reagent for efficient genome editing
Use the leading technology to deliver your Cas9 protein and your gRNA!
jetMESSENGER®- RNA transfection reagent for high genome editing
Take your CRISPR genome editing to the next level by using mRNA Cas9 and gRNA!
jetPRIME®- DNA transfection reagent for successful genome editing
Perform genome editing with plasmid(s) encoding Cas9 protein and gRNA!
Learn more about Polyplus transfection reagents range