jetPRIME®- DNA transfection reagent for successful genome editing
jetPRIME® is a powerful and versatile DNA and/or siRNA transfection reagent designed to ensure high nucleic acid transfection efficiency and excellent gene silencing in a variety of adherent cells.
- High DNA transfection efficiency for high genome editing
- Low amounts of nucleic acid
- Superior cell viability
- Easy to use
- Cost-effective CRISPR/Cas9 approach
jetPRIME® is very efficient in delivering DNA plasmid encoding Cas9 and gRNA, using little amount of DNA while respecting cell morphology and viability. Here is an example of a CRISPR/Cas9 plasmid successfully delivered by jetPRIME®.
See below Schematic map of the pCMV-Cas9-gRNA plasmid: