Dissolution Testing
Dissolution testing usually includes a time consuming step of solution preparation. The J.T.Baker® range of DILUT-IT™ solutions are pre-weighed and only need to be made up with purified water to create a ready to use dissolution media, reducing the average preparation time by more than 75%.
Produktauswahl