Multiple well cell culture plates, treated
Lieferant: TPP
92006 92012 92024 92096 92097 92697 92412 92406 92424 92696 92448 392012 92048 392006 392024 392096
TPPA92006EA 412 CHF
TPPA92006 TPPA92012 TPPA92024 TPPA92096 TPPA92097 TPPA92697 TPPA92412 TPPA92406 TPPA92424 TPPA92696 TPPA92448 TPPA392012 TPPA92048 TPPA392006 TPPA392024 TPPA392096
Multiple well cell culture plates, treated
Mikrotiterplatten Zellkultur-Platten
Clear PS, sterile cell culture dishes with lid.
  • Shaped corner on the plate and lid prevent incorrect realignment
  • Individual well rings in lid prevent cross-contamination
  • Ventilation feature on lid and plate maximises external/internal gas exchange and reduces media evaporation without altering CO₂ diffusion
  • 96 well plates are standardised with an interleaved barcode
  • Two well geometries - flat (F) and round (U) bottom
Designed for easy and stable stacking, these multiwell plates are characterised by yellow strip marking areas and easy-to-read, black embossed, alphanumeric well labelling. The cell growth surface treatment is limited to the well bottoms and offers optimal attachment and growth for a variety of cell types. The untreated well walls help flatten the liquid meniscus that forms, thereby reducing light diffusion. This feature, along with the optical clarity of the well bottoms, makes these plates ideal for microscopic examination and spectroscopic measurements.
The Triple Bagged (3-B) Packaging system meets strict cleanroom standards, preventing contamination from entering highly sensitive production areas. Each product is packaged individually in a tear-open bag. Each bag is labeled with a security number: 3 = external, 2 = middle, 1 = innermost bag. The innermost bag, Bag 1, is labeled with the product details, ie. product number, product description, lot number. The peel-open bags are increasingly sized, so that during handling of one bag the next bag in is not accidentally damaged rendering it contaminated. This significantly increases safe handling of substances in a sterile environment.
Zertifizierungen: Manufactured according to ANSI/SLAS 1-2204 to ANSI/SLAS 4-2004.
Lieferumfang: Supplied either individually wrapped or 4/6 plates per bag.