Kjeldahl digestion system, K-438
Lieferant: Büchi
458-0022EA 0 CHF
458-0022
Kjeldahl digestion system, K-438
Aufschlusssysteme Aufschluss-Systeme
The model K-438 is additionally equipped with an automatically controllable lift for the raising and lowering of sample tubes. This provides a completely automatic digestion. The control software provides two versions: a manual entry of digestion time and temperature, followed by an immediate start or programming temperature and time with the opportunity for a delayed start.
  • Fully automated digestion with choice of delayed start
  • Temperature profile with four temperature steps
Lieferumfang: Complete and ready to use, supplied with 20 digestion tubes, one 20 place sample rack, 1 suction module, mains cable and operating instructions.