Tiegelzangen
Lieferant: Nickel-Electro
231-0356EA 83.7 CHF
231-0356
Tiegelzangen
Zangen
Pure nickel (99,6%). For easy, safe handling of crucibles made from a wide variety of materials.
  • With grooved tips
  • Excellent corrosion resistance
  • Good thermal and electrical conductivity
  • Excellent thermal resistance