U-46619 ≥98%
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Prostaglandin H₂/thromboxane A₂ receptor agonist; stable analog of thromboxane A₂. Induces platelet aggregation at 32 nM and activates p38 MAPK.
Soluble in 100% ethanol, DMSO, dimethyl formamide (100 mg/ml) or PBS, pH 7.2 (2 mg/ml).
Formel:
C₂₁H₃₄O₄ Lager Temperatur: Gefrierschrank |
CAS-Nummer:
56985-40-1 |
|
Spezifikation Testergebnisse
Purity | ≥98% (HPLC-ELSD) |
Formulation | Pale yellow oil |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...