6-ECDCA ≥97% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyms: Obeticholic acid, (R)-4-((3R,5S,6R,7R,8S,9S,10S,13R,14S,17R)-6-ethyl-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, INT-747
BMLGR2460005 BMLGR2460025
ENZOBMLGR2460005EA 129 CHF
ENZOBMLGR2460005 ENZOBMLGR2460025
6-ECDCA ≥97% (durch TLC)
6-ECDCA
FXR ligand
A novel derivative of cholic acid which acts as a potent and selective FXR agonist (EC50= 99nM) displaying anticholeretic activity in an in vivo rat model of cholestasis. It not only inhibits vascular smooth muscle cell inflammation and migration but also promotes adipocyte differentiation and regulates adipose cell function in vivo.
Formel: C₂₆H₄₄O₄
MW: 420,63 g/mol
MDL Number: MFCD16621104
CAS-Nummer: 459789-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4472d076f5eb36858cf680297673807a2ac134ab&vid=7a498156c6740d90a6b3f3b328831b6df0bba528&prid=web&clid=SAPDAM