Palmityltrifluoromethylketone ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
BMLST3360010 BMLST3360050
ENZOBMLST3360010EA 102 CHF
ENZOBMLST3360010 ENZOBMLST3360050
Palmityltrifluoromethylketone ≥98% (durch TLC)
Palmityltrifluoromethylketone
Inhibitor of both Ca2+-dependent cytosolic cPLA2 (IC50=45 µM) and Ca2+-independent phospholipases A2 iPLA2 (IC50=3.8 µM).
  • Phospholipase inhibitor
White to off-white solid.
Soluble in 100% ethanol, dimethyl formamide or DMSO (warm); sparingly soluble in neutral buffers.
Formel: C₁₇H₃₁F₃O
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00797669
CAS-Nummer: 141022-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d657b687186ba2e9e2a3b024227c0d02fcb1a3f2&vid=8012a7d1c45d6b7e6ab3b4490f020bb14ba45279&prid=web&clid=SAPDAM