Fibronectin (human) ELISA kit
Lieferant: AbFrontier
Fibronectins bind cell surfaces and various compounds including collagen, fibrin, heparin, DNA, and actin. Fibronectins are involved in cell adhesion, cell motility, opsonization, wound healing, and maintenance of cell shape.
- Sensitivity: <0,910 ng/ml
- Form: Pre-coated plate
- Detection range: 1,25~80 ng/ml
Anastellin binds fibronectin and induces fibril formation. This fibronectin polymer, named superfibronectin, exhibits enhanced adhesive properties. Both anastellin and superfibronectin inhibit tumor growth, angiogenesis and metastasis. Anastellin activates p38 MAPK and inhibits lysophospholipid signaling.
Application: sELISA.
Sample type: Human serum, plasma, cell lysate, cultuer supernatant, buffered solution.
All kit components of this kit are stable at 2 to 8 °C. Any unused reconstituted standard should be discarded or frozen at –70 °C. Standard can be frozen and thawed one time only without loss of immunoreactivity.
Lieferumfang: Kit includes 96-well plate, washing buffer, standard protein, standard/sample dilution buffer, secondary antibody, anti-rabbit IgG HRP(X100), secondary antibody/anti-rabbit IgG HRP dilution buffer, substrate (TMB), stop solution, plate sealers and protocol booklet.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...