DNA Ladder 100 to 10000 bp, AMPIGENE®
Lieferant: Enzo Life Sciences
AMPIGENE®
ALEXENZGEN1040100EA 140 CHF
ALEXENZGEN1040100 ENZOENZGEN1040100
DNA Ladder 100 to 10000 bp, AMPIGENE®
Elektrophorese-Reagenzien Molekulargewichtsmarker
AMPIGENE® DNA ladder contains DNA fragments ranging from 100 bp to 10000 bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 500 bp, 1500 bp and 3000 bp to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5 µl load, for determining the approximate mass of DNA in samples with comparable intensity and sizes.
  • Ready to use format can be loaded straight onto a gel
  • Evenly spaced bands with intensified reference bands
  • Stable at room temperature
Lieferumfang: Kit contains ready to use AMPIGENE® DNA ladder 100 – 10000 bp and sample loading buffer.