Human recombinant FAP
Lieferant: Enzo Life Sciences
ALEXBML-SE409-0010EA 710 CHF
ALEXBML-SE409-0010 ENZOBMLSE4090010
Human recombinant FAP
Proteine und Peptide
Produced in a baculovirus expression system. Active recombinant human FAP (aa 29-760), representing a naturally-occurring cleaved (soluble) form.