HSP90α (human), ELISA Kit
Lieferant: Enzo Life Sciences
ENZOADIEKS895EA 1290 CHF
ENZOADIEKS895 ALEXADI-EKS-895
HSP90α (human), ELISA Kit
Tests ELISAs
Most sensitive, rapid ELISA kit for highly specific, quantitative detection of human HSP90α.
For the quantitative determination of HSP90α in cell lysates, serum, and tissue of human origin.
The HSP90α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Bestellinformation: Contains Microtiter plate, 5X Extraction reagent 2, Standard, Sample diluent, Wash buffer concentrate, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.