Anti-UBA6 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
BMLPW05250025 BMLPW05250100
ENZOBMLPW05250025EA 126 CHF
ENZOBMLPW05250025 ENZOBMLPW05250100
Anti-UBA6 Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For Western Blot

Recognizes human UBA6.

Type: Primary
Antigen: UBA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR