Anti-Y Mouse Monoclonal Antibody
Lieferant: Enzo Life Sciences
ALEXALX804174C100EA 460 CHF
ALEXALX804174C100 ENZOALX804174C100
Anti-Y Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1
Epitope: ...DIpYAE...Recognizes phosphotyrosine in the context of the surrounding amino acids, tolerating hydrophobic amino acids directly neighbouring the phosphotyrosine.
For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Reconstitute with 1ml water (15 minutes at room temperature).
Bestellinformation: Positive control included.
Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Many Species