Lauroyl peroxide zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

8.43861.0100EA 64.9 CHF
8.43861.0100
Lauroyl peroxide zur Synthese, Sigma-Aldrich®
Lauroyl peroxide
Formel: (C₁₁H₂₃CO)₂O₂
MW: 398,628 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008964
CAS-Nummer: 105-74-8
EINECS: 203-326-3
UN: 3106
ADR: 5.2,

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) slightly soluble
Melting Point 49 °C
Molar Mass 398.63 g/mol
Density 0.91 g/cm3 (20 °C)
Ignition Point 112 °C
HS Code 2915 90 70
Density 0,91 g/cm3 (20 °C)
Ignition temperature 112 °C
Melting Point 49 °C
Solubility (20 °C) slightly soluble
Storage temperature 15...25 °C
Assay (iodometric) ≥98%
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR