Anti-ACTB Rabbit Polyclonal Antibody

Lieferant: AbFrontier
LF-PA0209A LF-PA0209
ABFRLF-PA0209AEA 560 CHF
ABFRLF-PA0209A ABFRLF-PA0209
Anti-ACTB Rabbit Polyclonal Antibody
Antikörper
  • Western Blot: 1:10,000;

Type: Primary
Antigen: ACTB (actin, beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR