Sterilisations-Autoklaven
Lieferant: SANOCLAV WOLF
01005 01002 01001 01033 01032
481-4639EA 10950 CHF
481-4639 481-4640 481-0264 SANC01033 SANC01032
Sterilisations-Autoklaven
Sterilisatoren Autoklaven (Dampfsterilisatoren)
Für die Sterilisation im gespannten Dampf und Wasser
  • Equipped with safety valve, overheat protection, drain valve and manometer
  • Pressure chamber with stainless steel lid and bowl
  • Microprocessor control
  • Filter system for exhaust air filtration
RS 232 interface with program for recording time and temperature on an external PC. Flexible PT 100 sensor for liquids (with the media sensor switched on, a pre- and post-heating temperature of 60 °C is maintained).
Safety interlock in La-MCS or La-VA-MCS, temperature-dependent at over 97 °C or locked at over 8More Product Information