Autoclaves, Priorclave Compact
Lieferant: Priorclave
B40-SMART B60-SMART V60-SMART V60-BASE B40-BASE B60-BASE
PRIOB40-SMARTEA 13780 CHF
PRIOB40-SMART PRIOB60-SMART PRIOV60-SMART PRIOV60-BASE PRIOB40-BASE PRIOB60-BASE
Autoclaves, Priorclave Compact
Sterilisatoren Autoklaven (Dampfsterilisatoren)
Compact 40 benchtop and Compact 60 floor standing or benchtop autoclaves for the low volume user, with flexible operation usually only available in larger autoclave units. This flexibility is delivered by the TACTROL™ microprocessor control system, specially developed for laboratory autoclaves, with programming ranging from simple cycles to fully featured multi-program operation with logged records. All Priorclaves come complete with Biomaster antimicrobial coating, helping improved laboratory hygiene; it is effective against all bacteria and fungi including MRSA and therefore helps minimiseMore Product Information