Dissecting trays
Lieferant: NICKEL ELECTRO
625 626
NICK625EA 105 CHF
NICK625 NICK626
Dissecting trays
Schalen/Schüsseln/Töpfe Präparierwannen
Dissecting trays made of aluminium.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean
Order Now
TECHNISCHE DATEN
Environmentally Preferable