Human recombinant FGF-8

Lieferant: ENZO LIFE SCIENCES

ENZPRT1740025
ENZOENZPRT1740025EA 359 CHF
ENZOENZPRT1740025
Human recombinant FGF-8
Proteine und Peptide

Produced in E. coli.

New Product

Order Now

TECHNISCHE DATEN

Protein/Peptid-Name FGF-8
Environmentally Preferable
Proteinsynonym Fibroblast growth factor-8|AIGF|FGF-8b|HBGF-8
Protein/Peptid-Typ Rekombinant
Spezies Human
Ursprung
Purity >95% (SDS and HPLC)
Zusammensetzung Lyophilized from 10 mM sodium phosphate, pH 8,0, containing 75 mM NaCl.
Molecular weight ~22,5 kDa
Reinigung Sterile filtered through a 0,2 micron filter.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR