Sertraline hydrochloride ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyms: (1S,4S)-1-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-4-(methylamino)naphthalene Hydrochloride

BMLNS1150010
ENZOBMLNS1150010EA 102 CHF
ENZOBMLNS1150010
Sertraline hydrochloride ≥98% (durch TLC)
Sertraline hydrochloride

Serotonin transporter inhibitor


A selective serotonin transporter inhibitor. In clinical use as an antidepressant. Displays antiproliferative activity on human colorectal carcinoma cells.

Formel: C₁₇H₁₇Cl₂N
MDL Number: MFCD00895772
CAS-Nummer: 79559-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee1137c05dbbbde1c18dd6f957525475ae8c6dbc&vid=817fd8a8883f6210d68510a6421b3c683c5cb13b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR