Levocabastine HCl ≥99% (durch HPLC), weißer Feststoff

Lieferant: ENZO LIFE SCIENCES

BMLNH1070010
ENZOBMLNH1070010EA 295 CHF
ENZOBMLNH1070010
Levocabastine HCl ≥99% (durch HPLC), weißer Feststoff
Levocabastine HCl

Histamine H₁ receptor antagonist. An antagonist of low affinity neurotensin receptors.


Soluble in DMSO, methanol or DMF.

Formel: C₂₆H₂₉FN₂O₂·HCl
Lager Temperatur: Raumtemperatur
CAS-Nummer: 79547-78-7

Order Now

Spezifikation Testergebnisse

Purity ≥99% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR