Sulindac sulphone ≥94% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyms: Z-5-Fluoro-2-methyl-1-[p-(methyl-sulphonyl)benzylidene]indene-3-acetic acid

BMLAP2010025
ENZOBMLAP2010025EA 328 CHF
ENZOBMLAP2010025
Sulindac sulphone ≥94% (durch HPLC)
Sulindac sulphone

Induces apoptosis but does not inhibit cyclooxygenase. Inhibits growth and induces apoptosis in human cancer cell lines. Metabolite of Sulindac.


  • Sulindac metabolite


Yellow solid.


Soluble in DMSO (>25mg/ml) or 100% ethanol (3mg/ml, warm).

Formel: C₂₀H₁₇FO₄S
MW: 372.4 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00871143
CAS-Nummer: 59864-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR