Ascomycin (aus Streptomyces species) ≥90% (durch HPLC, TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyms: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-8-Ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone

BMLA1650001
ENZOBMLA1650001EA 268 CHF
ENZOBMLA1650001
Ascomycin (aus Streptomyces species) ≥90% (durch HPLC, TLC)
Ascomycin

Ascomycin is a close structural analog of FK506 and is a potent immunosuppressant. It binds to FKBP (FK506-binding protein) and suppresses its proline rotamase activity. The ascomycin-FKBP complex inhibits calcineurin, a type 2B phosphatase.


Off-white solid


Soluble in DMSO, methanol or ethanol


Verpackung: Shipping temperature: Ambient

Formel: C₄₃H₆₉NO₁₂
MW: 790 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD06198665
CAS-Nummer: 104987-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR