Anti-TLR6 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: ENZO LIFE SCIENCES
ALX804363FC100
ENZOALX804363FC100EA 796 CHF
ENZOALX804363FC100
Anti-TLR6 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Host: Mouse, Isotype: IgG1

Recognizes human TLR6.

Type: Primary
Antigen: TLR6
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR