Cyclothiazide

Lieferant: ENZO LIFE SCIENCES

Synonyms: 6-Chloro-3,4-dihydro-3-(2-norbornen-5-yl)-2H-1,2-4-benzothiadiazine-7-sulphonamide 1,1-Dioxide, Cyclothiazide (mixture of isomers)

ALX550338M050
ENZOALX550338M050EA 408 CHF
ENZOALX550338M050
Cyclothiazide
Cyclothiazide

AMPA receptor activator


Potently inhibits the desensitization of AMPA receptors. Antihypertensive.

Formel: C₁₄H₁₆ClN₃O₄S₂
MW: 389,87 g/mol
MDL Number: MFCD00210192
CAS-Nummer: 2259-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR