Serotonin Hydrochlorid

Lieferant: ENZO LIFE SCIENCES

Synonyms: 5-Hydroxytryptamine Hydrochloride

ALX550328G001 ALX550328M050
ENZOALX550328G001EA 371 CHF
ENZOALX550328G001 ENZOALX550328M050
Serotonin Hydrochlorid
Serotonin Hydrochlorid
Formel: C₁₀H₁₃ClN₂O
MDL Number: MFCD00012686
CAS-Nummer: 153-98-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR