Necrostatin-5 ≥98% (durch NMR)

Lieferant: ENZO LIFE SCIENCES

Synonyms: 2-((3-(4-Methoxyphenyl)-4-oxo-3,4,5,6,7,8-hexahydrobenzo[4,5]thieno[2,3-d]pyrimidin-2-yl)thio)acetonitrile

ALX430169M005 ALX430169M025
ENZOALX430169M005EA 78.6 CHF
ENZOALX430169M005 ENZOALX430169M025
Necrostatin-5 ≥98% (durch NMR)
Necrostatin-5

Necroptosis inhibitor


Necroptosis inhibitor. In contrary to the mechanism of necrostatin-1 (Prod.No. BML-AP309) it was shown that necrostatin-5 inhibits RIP1 kinase indirectly. A potent inhibitor of immunoprecipitated, but not recombinant RIP1.

Formel: C₁₉H₁₇N₃O₂S₂
CAS-Nummer: 337349-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR