R-sulphoraphane ≥98% (durch UPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyms: (R)-1-Isothiocyanato-4-(methylsulphinyl)butane, L-sulphoraphane

ALX350230M010
ENZOALX350230M010EA 512 CHF
ENZOALX350230M010
R-sulphoraphane ≥98% (durch UPLC)
R-sulphoraphane

Carciogenesis inhibitor


Potent, selective inducer of phase II detoxification enzymes. Inhibits chemically induced mammary tumor formation in rats.


Chiral natural product isolated from broccoli.

Formel: C₆H₁₁NOS₂
MW: 177.2876 g/mol
MDL Number: MFCD09039283
CAS-Nummer: 142825-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR