Okadasäure

Lieferant: ENZO LIFE SCIENCES

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA

ALX350003C025 ALX350003C050 ALX350003C100 ALX350003M001
ENZOALX350003C025EA 163 CHF
ENZOALX350003C025 ENZOALX350003C050 ENZOALX350003C100 ENZOALX350003M001
Okadasäure
Okadaic acid
Formel: C₄₄H₆₈O₁₃
MW: 805.02 g/mol
MDL Number: MFCD00083455
CAS-Nummer: 78111-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR