Dithiothreitol (DTT, Cleland's Reagenz)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

ALX280001G005 ALX280001G010 ALX280001G025 ALX280001G050
ENZOALX280001G005EA 128 CHF
ENZOALX280001G005 ENZOALX280001G010 ENZOALX280001G025 ENZOALX280001G050
Dithiothreitol (DTT, Cleland's Reagenz)
Dithiothreitol (DTT, Cleland's Reagenz)
Formel: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Siedepunkt: ∼125 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash-Pt: >110 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00004877
CAS-Nummer: 3483-12-3
EINECS: 222-468-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR