Z-VEID-FMK
Lieferant: ENZO LIFE SCIENCES
ALX260143R020 ALX260143R100
ENZOALX260143R020EA 322 CHF
ENZOALX260143R020 ENZOALX260143R100
Z-VEID-FMK
Proteine und Peptide
Caspase-6 inhibitor
Irreversible, cell permeable inhibitor of caspase-6. The inhibitor is designed as a methyl ester to facilitate cell permeability.