Anti-CASP9 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX210815C100
ENZOALX210815C100EA 715 CHF
ENZOALX210815C100
Anti-CASP9 Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For Western Blot

Recognizes human, mouse, rat, hamster, rabbit, pig, dog, monkey and bovine caspase-9. Detects procaspase-9 and a caspase-9 cleavage product by Western blot.

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR