PGE2 CLIA kit
Lieferant: ENZO LIFE SCIENCES
ADI910001
ENZOADI910001EA 695 CHF
ENZOADI910001
PGE2 CLIA kit
Tests ELISAs
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.
The PGE2 CLIA kit is a sensitive chemiluminescent competitive enzyme immunoassay kit for the quantitative determination of PGE2 in biological fluids from any species.
Bestellinformation: Contains White GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, CLIA Substrate.