Anti-Opioid Receptor kappa Rabbit Polyclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ADI905799100
ENZOADI905799100EA 526 CHF
ENZOADI905799100
Anti-Opioid Receptor kappa Rabbit Polyclonal Antibody
Antikörper
The opioid receptors delta (DOP, OP1), kappa (KOP, OP2), and mu (MOP, OP3) are members of the seven-transmembrane domain-containing receptor family of G-protein coupled receptors, sharing nearly 60% identity, but varying greatly in the extracellular N-terminus region of the receptors. The delta, kappa and mu-receptors serve as functional receptors for endogenous enkephalin, dynorphin, and beta-endorphin peptide ligands, respectively, mediating their signal via coupling to Gi/o alpha subunits to inhibit adenylyl cyclases and calcium conductance (N- and L-type channels), and to stimulate MAPKMore Product Information