Lauroyl peroxide 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Dilauroyl peroxide

349741000 349745000
ACRO349741000EA 81.3 CHF
ACRO349741000 ACRO349745000
Lauroyl peroxide 99%
Lauroyl peroxide

CAS No.: 105-74-8

Formel: (C₁₁H₂₃CO)₂O₂
MW: 398.628 g/mol
Melting Pt: 53…57 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00008964
CAS-Nummer: 105-74-8
UN: 3106
ADR: 5.2

Order Now

Spezifikation Testergebnisse

Appearance White Powder or granules or flakes or pellets
Infrared spectrum Conforms
Melting point 53 to 57 °C
Titration Iodometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR