Anti-PRDX1 Rabbit Polyclonal Antibody
Lieferant: AbFrontier
LF-PA0095
ABFRLF-PA0095EA 311 CHF
ABFRLF-PA0095
Anti-PRDX1 Rabbit Polyclonal Antibody
Antikörper
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, particularly amino-terminal region containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1- Cys Prx subgroup, the otherMore Product Information