Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 36H1]
Lieferant: AbFrontier
LF-MA0275
ABFRLF-MA0275EA 311 CHF
ABFRLF-MA0275
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 36H1]
Antikörper
Carcinoembryonic antigen (CEA) was first described in 1965 by Gold and Freedman when they identified an antigen that was present in both fetal colon and colon adenocarcinoma but that appeared to be absent from
healthy adult colon. CEA is a glycoprotein containing approximately 50% carbohydrate with a molecular weight of 180 kDa.
CEA and related genes make up the CEA family belonging to the immunoglobulin superfamily. In humans, the carcinoembryonic antigen family consists of 29 genes, 18 of which are normally expressed.
CEA has proven to be a suitable target antigen for the detection ofMore Product Information