Anti-PLK1 Mouse Monoclonal Antibody [clone: 18D4]
Lieferant: AbFrontier
LF-MA0194
ABFRLF-MA0194EA 311 CHF
ABFRLF-MA0194
Anti-PLK1 Mouse Monoclonal Antibody [clone: 18D4]
Antikörper
Polo-like kinases (Plks) are important regulators of the cell cycle. Plk1, the most studied Plk, has been implicated in regulating centrosome maturation, mitotic entry, sister chromatid cohesion, the anaphase-promoting complex/cyclosome (APC/C), and cytokinesis. Sister chromatid separation and the subsequent formation of two genetically identical daughter cells depend on the symmetrical attachment of all chromosomes to the mitotic spindle, a process called chromosome biorientation. Several mitosis-specific protein kinases have been implicated in bipolar spindle assembly and chromosomeMore Product Information