Anti-HSPA13 Mouse Monoclonal Antibody [clone: 16A12]

Lieferant: AbFrontier
LF-MA0101
ABFRLF-MA0101EA 311 CHF
ABFRLF-MA0101
Anti-HSPA13 Mouse Monoclonal Antibody [clone: 16A12]
Antikörper
The Heat shock protein 70(HSP70) family was found in many intracellular compartments. Members of this protein occur in chloroplasts, endoplasmic reticulum, mitochondria, and cytosol. These proteins are induced by a variety of biological stresses, including heat stress, in every organism. HSP70 serves a variety of roles: 1) It acts as molecular chaperones facilitating the assembly of multi-protein complexes, 2) It participates in the translocation of polypeptides across cell membranes and to the nucleus 3) It aids in the proper folding of nascent polypeptide chains. HSP70 is mitochondrialMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol HSPA1A
Environmentally Preferable
Antigen-Name Heat shock 70kDA protein 1A
Antigen-Synonyme HSPA1|HSP70-1A|HSP72|HSP70-1|HSP70.1|HEL-S-103|HSP70I
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Unconjugated
Klon 16A12
Reaktivität Human
Wirt Mouse
Gen-ID 3303
Isotyp IgG2b
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunpräzipitation Yes
Immunogen Recombinant Human HSP70 protein purified E.coli
Reinigung Ammonium sulfate precipitation
Kreuzadsorption none
Lagertemperatur Store for 1 year at –20 °C from date of shipment
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR