Anti-TXNRD1 Mouse Monoclonal Antibody [clone: 5A5]
Lieferant: AbFrontier
LF-MA0020
ABFRLF-MA0020EA 311 CHF
ABFRLF-MA0020
Anti-TXNRD1 Mouse Monoclonal Antibody [clone: 5A5]
Antikörper
The mammalian thioredoxin reductases (TrxRs) are a family of selenocysteine-containing pyridine nucleotide-disulfide oxido-reductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate selenocysteine (SeCys) residue in the conserved sequence(-Gly-Cys-SeCys-Gly). TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redoxMore Product Information