Strip Plates, 96-Well
Lieferant: Greiner Bio-One
756070_100 762070_100 701071_100 705070_100 705071_100 705073_100 705074_100 705075_100 705076_100 756071_100 762071_100 705185_400 762001_400 767001_400 762074_100 762077_100 762075_100 762076_100 754101_400
738-0041EA 523 CHF
738-0041 738-0043 738-0037 738-0039 738-0038 737-0176 737-0177 737-0178 737-0179 738-0042 738-0044 737-0189 737-0194 737-0196 738-0045 738-0048 738-0046 738-0047 737-0190
Strip Plates, 96-Well
Mikrotiterplatten Immunologie-Mikrotiterplatten
PS, without lid. Microplates in strip format offer the advantage of greater flexibility. Individual strips can be removed from the support frame so that the number of tests to be performed can be adjusted to the number of samples, and is not predetermined by the microplate format used. Strip plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600), medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200), black (FLUOTRAC™) or white (LUMITRAC™)More Product Information