PCR/qPCR Instruments
Featured Solutions
The VWR® Collection PCR thermal cycler XT96 combines industry-inspired innovation with the reliable quality of German manufacturing - housing both in a compact, low-noise ventilation design that fits on virtually any benchtop.
Additional Resources
The AriaMx Real-Time PCR system is a fully integrated quantitative PCR amplification, detection and data analysis system…
Q qPCR cycler - A faster, smaller, better way to qPCR
Q uses a patented magnetic induction technology to rapidly heat and cool samples to acquire data in as little as 25 minutes.