Blood and tissue genomic DNA isolation
Mag-BIND® Blood & Tissue DNA HDQ 96 Kit
Mag-BIND® Blood & Tissue DNA HDQ 96 Kit offers a rapid, high throughput, cost-effective solution for purifying high quality genomic DNA from from 100 to 250 µl of blood samples: (we have also solutions for volumes as large as 10 ml), saliva, swabs, mouse tails, dried blood spots, tissues, or 5x10^6 cultured cells. Mag-BIND® Particles HDQ provide quick magnetic response times thereby reducing overall processing time.
The kit is automatable on most open-ended automated platforms and is very time-efficient as 384 samples can be processed in 2 hours. This system combines the reversible nucleic acid-binding properties of Mag-BIND® paramagnetic particles with the proven efficiency of Omega Bio-tek’s buffer chemistry to provide a rapid and robust method to isolate DNA from a variety of biological samples. The system yields high quality DNA that is suitable for direct use in most downstream applications, such as amplification, NGS, and enzymatic reactions.
- Rapid - Process 384 samples in 2 hours
- Bead-based - Scalable DNA purification; 100 to 250 µl input
(we have solutions for volumes of blood of up to 10 ml) - Quality - High quality DNA suitable for qPCR, NGS, etc
- Automatable - Adaptable on most open-ended liquid handlers
- Cost-effective - On average 60% less than the competition
Request your free sample |
Scientific publication list |