Upstream processing for vaccines
Optimal production of vaccines is achieved in mammalian expression and insect systems using cell lines that are anchorage-dependent or suspension enabled.
For most vaccines, optimal production is achieved in mammalian expression and insect systems using cell lines that are anchorage-dependent or suspension enabled.
Avantor, delivered by VWR provides culture systems that support processes development, scale-up, and full manufacturing. Our systems include a wide variety of:
- Culture flasks and shaker platforms
- Roller bottles and roller bottle racks
- Sterile cell factories
- Microcarriers to allow culture of cell lines in larger bioreactors
For expansion of your production cell line, we offer media components with enriched growth supplements so that you can design your own formulations. To support cell growth, we offer attachment factors and animal sera to enhance attachment. For mRNA vaccines manufacturing workflows using cell-free synthesis, enzymes and nucleotides are available.
Avantor J.T.Baker® brand offers the only cGMP manufactured IPTG to ICHQ7 standards, providing biopharma researchers and manufacturers with a solution to help scale novel vaccines from research and development to clinical then full scale manufacturing with speed, quality and confidence.
Explore solutions for upstream processing of vaccines
Product choices for discovery, product development & pre-clinical
Product choices for scale-up & production
Ready to Get Started?
Ready to Maximize Production Efficiency?
Our Bioprocessing Solutions team can help you streamline your biomanufacturing processes!