Human PEDF ELISA Kit
Lieferant: Abcam
AB246535-1X96 AB246535-10X96
ABCAAB246535-1X96EA 944 CHF
ABCAAB246535-1X96 ABCAAB246535-10X96
Human PEDF ELISA Kit
Tests ELISAs
Human PEDF ELISA kit is a single-wash 90-min Simplestep used to quantify Human PEDF with a sensitivity of 146 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders.