Human AMH ELISA Kit
Lieferant: Abcam
AB318930-10X96 AB318930-1X384 AB318930-96TST
ABCAAB318930-10X96EA 7740 CHF
ABCAAB318930-10X96 ABCAAB318930-1X384 ABCAAB318930-96TST
Human AMH ELISA Kit
Tests ELISAs
Human AMH ELISA kit is a single-wash 90-min Simplestep used to quantify Human AMH with a sensitivity of 18,389 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput.