Human AMH ELISA Kit
Lieferant: Abcam
Human AMH ELISA kit is a single-wash 90-min Simplestep used to quantify Human AMH with a sensitivity of 18,389 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput.