Rat IFN gamma ELISA Kit
Lieferant: Abcam
AB46107-96
AB46107-2X96
ABCAAB46107-96EA
938
CHF
ABCAAB46107-96
ABCAAB46107-2X96
Rat IFN gamma ELISA Kit
Tests
ELISAs
Rat IFN gamma ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat IFN gamma in Rat in Cell culture supernatant, Serum samples.