Rat IFN gamma ELISA Kit
Lieferant: Abcam
AB46107-96 AB46107-2X96
ABCAAB46107-96EA 938 CHF
ABCAAB46107-96 ABCAAB46107-2X96
Rat IFN gamma ELISA Kit
Tests ELISAs
Rat IFN gamma ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat IFN gamma in Rat in Cell culture supernatant, Serum samples.